Assay Details
| Gene Symbol: ZFYVE27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011539254.2 | XP_011537556.1 | 2 - 3 | 370 | 63 | |
| XM_011539255.2 | XP_011537557.1 | 2 - 3 | 377 | 63 | ||
| XM_017015650.1 | XP_016871139.1 | 2 - 3 | 370 | 63 | ||
| GenBank mRNA | AK057481.1 | - | 2 - 3 | 377 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK057481.1, XM_011539254.2, XM_011539255.2, XM_017015650.1 transcripts.
Set Membership: |
|

ZFYVE27
Genomic Map