Assay Details
| Gene Symbol: ASB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198243.2 | NP_937886.1 | 6 | 3349 | 74 | |
| GenBank mRNA | AK057921.1 | - | 1 | 252 | 74 | |
| AK094123.1 | - | 1 | 1606 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000967.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000561192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000561192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000561192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000561192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | ENST00000561192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008967.hg.1 | ENST00000332783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008967.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008967.hg.1 | TC15000967.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500161909.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500161909.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500161909.hg.1 | TC1500008545.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500161909.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500161909.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000332783.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000343276.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000558747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000561192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000561192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000561192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000561192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | ENST00000561192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_024708.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_198243.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008545.hg.1 | NM_198243.2 | Refseq |

ASB7
Genomic Map