Assay Details
| Gene Symbol: PACRG-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110871.1 | 2 - 3 | 1006 | 103 | ||
| GenBank mRNA | AK058177.1 | - | 2 - 3 | 1006 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013812.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195289.hg.1 | TC0600013812.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195294.hg.1 | TC0600013812.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600195286.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195286.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195286.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195286.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195286.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195286.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195286.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195286.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195286.hg.1 | TC0600013812.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195287.hg.1 | TC0600013812.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | NR_110872.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195295.hg.1 | TC0600013812.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | ENST00000418665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | ENST00000433489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | ENST00000440924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | NR_110871.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600195290.hg.1 | TC0600013812.hg.1 | Clariom D TC |

PACRG-AS2
Genomic Map