| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010934.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010934.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010934.hg.1 |
TC01000674.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010935.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010935.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01010935.hg.1 |
TC01000674.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100159927.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159927.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159927.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159927.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159927.hg.1 |
TC0100008416.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100159928.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159928.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159928.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159928.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159928.hg.1 |
TC0100008416.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008416.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159926.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159926.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159926.hg.1 |
ENST00000371265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100159926.hg.1 |
NM_057176.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100159926.hg.1 |
TC0100008416.hg.1 |
Clariom D TC |