Assay Details
| Gene Symbol: IFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002169.2 | NP_002160.1 | 1 | 409 | 95 | |
| GenBank mRNA | BC093755.1 | - | 1 | 409 | 7939600 | 95 |
| BC093757.1 | - | 1 | 409 | 7939602 | 95 | |
| V00541.1 | - | 1 | 184 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

IFNA5
Genomic Map