| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000467617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000467617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000467617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000476110.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000476110.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000476110.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000476110.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
ENST00000476110.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008195.hg.1 |
ENST00000417406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008195.hg.1 |
ENST00000417406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000216225.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000467617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000467617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000467617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000476110.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000476110.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000476110.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000476110.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
ENST00000476110.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009278.hg.1 |
NM_014248.3 |
Refseq |