Assay Details
| Gene Symbol: PTPN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002833.3 | NP_002824.1 | 8 - 9 | 1576 | 75 | |
| GenBank mRNA | BC010863.1 | - | 8 - 9 | 1080 | 3895727 | 75 |
| BC071574.1 | - | 9 - 10 | 1497 | 5276190 | 75 | |
| BT007405.1 | - | 8 - 9 | 1061 | 75 | ||
| M83738.1 | - | 8 - 9 | 1569 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001667.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000561731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000561731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000561731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000561731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000563835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000563835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000563835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000564970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000564970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000564970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000568108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000568108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000568108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000568108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15016248.hg.1 | ENST00000306726.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15016248.hg.1 | ENST00000544966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15016248.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15016248.hg.1 | TC15001667.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000561731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000561731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000561731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000561731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000563835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000563835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000563835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000564970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000564970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000564970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000568108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000568108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000568108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000568108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010893.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500185451.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500185451.hg.1 | ENST00000563835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500185451.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500185451.hg.1 | NM_002833.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500185451.hg.1 | TC1500010893.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500185451.hg.1 | ENST00000306726.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500185451.hg.1 | ENST00000563835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500185451.hg.1 | ENST00000618819.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500185451.hg.1 | NM_002833.3 | Refseq |

PTPN9
Genomic Map