| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02043175.hg.1 |
ENST00000264276.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02043175.hg.1 |
ENST00000457679.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02043175.hg.1 |
NM_020919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02043175.hg.1 |
TC02002678.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200213346.hg.1 |
ENST00000264276.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200213346.hg.1 |
ENST00000439495.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200213346.hg.1 |
NM_020919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200213346.hg.1 |
ENST00000264276.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200213346.hg.1 |
ENST00000439495.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200213346.hg.1 |
NM_020919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200213346.hg.1 |
TC0200015434.hg.1 |
Clariom D TC |