Assay Details
| Gene Symbol: SETD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306199.1 | NP_001293128.1 | 5 - 6 | 1279 | 106 | |
| NM_030648.3 | NP_085151.1 | 5 - 6 | 1279 | 106 | ||
| XM_017008661.1 | XP_016864150.1 | 3 - 4 | 372 | 106 | ||
| GenBank mRNA | AB051504.1 | - | 5 - 6 | 914 | 106 | |
| AF448510.1 | - | 5 - 6 | 914 | 106 | ||
| AF462150.1 | - | 5 - 6 | 647 | 106 | ||
| AK310856.1 | - | 5 - 6 | 748 | 106 | ||
| BC121055.1 | - | 5 - 6 | 823 | 40118363 | 106 | |
| BC121056.1 | - | 5 - 6 | 823 | 40118365 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
cellular response to DNA damage stimulus
chromatin modification
peptidyl-lysine monomethylation
peptidyl-lysine dimethylation
histone lysine methylation
response to ethanol
positive regulation of transcription, DNA-templated
regulation of histone H3-K9 methylation
heterochromatin organization
p53 binding
chromatin binding
protein binding
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
transcription, DNA-templated
cellular response to DNA damage stimulus
chromatin modification
peptidyl-lysine monomethylation
peptidyl-lysine dimethylation
histone lysine methylation
response to ethanol
positive regulation of transcription, DNA-templated
regulation of histone H3-K9 methylation
heterochromatin organization
p53 binding
chromatin binding
protein binding
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000515101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000515101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | ENST00000515101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000404104.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000406354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000515101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000515101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | ENST00000515101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_001306200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012969.hg.1 | NR_131339.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | ENST00000515101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | ENST00000274031.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | ENST00000506866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | ENST00000515101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | NM_001306199.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | NM_030648.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191068.hg.1 | TC0400012969.hg.1 | Clariom D TC |

SETD7
Genomic Map