Assay Details
| Gene Symbol: NOSTRIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052946.3 | NP_443178.2 | 1 - 2 | 81 | 69 | |
| GenBank mRNA | BC014189.2 | - | 1 - 2 | 81 | 4650302 | 69 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC014189.2, NM_052946.3 transcripts.
Set Membership: |
|

NOSTRIN
Genomic Map