Assay Details
| Gene Symbol: CNNM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017649.4 | NP_060119.3 | 6 - 7 | 2400 | 97 | |
| GenBank mRNA | AF216962.1 | - | 6 - 7 | 1611 | 97 | |
| AK023066.1 | - | 6 - 7 | 1514 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000769.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369875.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369875.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369875.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369875.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369875.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000616802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | ENST00000616802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008758.hg.1 | NM_199077.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009155.hg.1 | ENST00000345419.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009155.hg.1 | ENST00000369878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009155.hg.1 | ENST00000433628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009155.hg.1 | ENST00000457502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009155.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009155.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009155.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009155.hg.1 | TC10000769.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159987.hg.1 | TC1000008758.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369875.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369875.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369875.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369875.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369875.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000369878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000433628.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000475511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000616802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | ENST00000616802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_017649.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199076.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199077.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008758.hg.1 | NM_199077.2 | Refseq |

CNNM2
Genomic Map