Assay Details
| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022834.4 | NP_073745.2 | 2 - 3 | 858 | 84 | |
| NM_199121.2 | NP_954572.2 | 2 - 3 | 463 | 84 | ||
| GenBank mRNA | AK075366.1 | - | 2 - 3 | 681 | 84 | |
| AK096773.1 | - | 2 - 3 | 270 | 84 | ||
| AK291557.1 | - | 2 - 3 | 665 | 84 | ||
| BC003543.1 | - | 1 - 2 | 142 | 84 | ||
| BC059409.1 | - | 2 - 3 | 665 | 30349211 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000040.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK096773.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01000473.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000473.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000473.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000473.hg.1 | TC01000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000474.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000474.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000474.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000474.hg.1 | TC01000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000468.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000468.hg.1 | ENST00000404702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000468.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000468.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000468.hg.1 | TC01000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000475.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000475.hg.1 | ENST00000404702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000475.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000475.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000475.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000475.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000475.hg.1 | TC01000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000469.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000469.hg.1 | ENST00000404702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000469.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000469.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000469.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000469.hg.1 | TC01000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000471.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000471.hg.1 | ENST00000404702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000471.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000471.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000471.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000471.hg.1 | TC01000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000477.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000477.hg.1 | ENST00000404702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000477.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000477.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000477.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000477.hg.1 | TC01000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000471398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000471398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000471398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144858.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144858.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144858.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144858.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144858.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144858.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144858.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144858.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144858.hg.1 | TC0100006516.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144864.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144864.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144864.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144864.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144864.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144864.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144864.hg.1 | TC0100006516.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144865.hg.1 | TC0100006516.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144867.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144867.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144867.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144867.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144867.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144867.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144867.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144867.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144867.hg.1 | TC0100006516.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144857.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144857.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144857.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144857.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144857.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144857.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144857.hg.1 | TC0100006516.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000471398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000471398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000471398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006516.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144868.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144868.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144868.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144868.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144868.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144868.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144868.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144868.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144868.hg.1 | TC0100006516.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | ENST00000471398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | ENST00000471398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144861.hg.1 | TC0100006516.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144863.hg.1 | ENST00000471398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144863.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144863.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144863.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144863.hg.1 | ENST00000471398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144863.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144863.hg.1 | ENST00000495558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144863.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144863.hg.1 | TC0100006516.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144869.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144869.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144869.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144869.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144869.hg.1 | ENST00000338660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144869.hg.1 | ENST00000476993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144869.hg.1 | NM_022834.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144869.hg.1 | NM_199121.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144869.hg.1 | TC0100006516.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 172273 | - | - |

VWA1
Genomic Map