Assay Details
| Gene Symbol: CDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030928.3 | NP_112190.2 | 2 - 3 | 406 | 58 | |
| GenBank mRNA | AB053172.1 | - | 2 - 3 | 406 | 58 | |
| AF321125.1 | - | 2 - 3 | 377 | 58 | ||
| BC000137.2 | - | 2 - 3 | 368 | 3048353 | 58 | |
| BC008676.1 | - | 2 - 3 | 244 | 58 | ||
| BC008860.2 | - | 2 - 3 | 299 | 58 | ||
| BC009410.1 | - | 2 - 3 | 362 | 3613566 | 58 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000686.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication checkpoint
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
DNA replication
regulation of DNA-dependent DNA replication initiation
regulation of nuclear cell cycle DNA replication
DNA binding
protein binding
DNA replication checkpoint
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
DNA replication
regulation of DNA-dependent DNA replication initiation
regulation of nuclear cell cycle DNA replication
DNA binding
protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000569140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000569140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000569140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000569140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | ENST00000569140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009681.hg.1 | ENST00000301019.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009681.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009681.hg.1 | TC16000686.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009682.hg.1 | ENST00000301019.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009682.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009682.hg.1 | TC16000686.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009683.hg.1 | ENST00000301019.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009683.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009683.hg.1 | TC16000686.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009679.hg.1 | ENST00000301019.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009679.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009679.hg.1 | TC16000686.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600165901.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165901.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165901.hg.1 | TC1600008845.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600165901.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165901.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165900.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165900.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165900.hg.1 | TC1600008845.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600165900.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165900.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165897.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165897.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165897.hg.1 | TC1600008845.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600165897.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165897.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165903.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165903.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165903.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165903.hg.1 | TC1600008845.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600165903.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165903.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165903.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000569140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000569140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000569140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000569140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | ENST00000569140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008845.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165902.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165902.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165902.hg.1 | NM_030928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165902.hg.1 | TC1600008845.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600165902.hg.1 | ENST00000301019.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165902.hg.1 | ENST00000562747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165902.hg.1 | NM_030928.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 160015 | - | - |

CDT1
Genomic Map