| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026009.hg.1 |
ENST00000304031.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026009.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026009.hg.1 |
TC02001625.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000406618.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000406618.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000406618.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000406618.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000406618.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000445102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000445102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000445102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000445102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000458740.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000458740.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000458740.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000458740.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000631166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
ENST00000631166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026010.hg.1 |
ENST00000304031.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026010.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026010.hg.1 |
TC02001625.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000402541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000406618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000406618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000406618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000406618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000406618.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000445102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000445102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000445102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000445102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000458740.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000458740.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000458740.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000458740.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000631166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
ENST00000631166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016640.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200225841.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225841.hg.1 |
ENST00000445102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225841.hg.1 |
ENST00000458740.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225841.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200225841.hg.1 |
ENST00000304031.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225841.hg.1 |
ENST00000445102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225841.hg.1 |
ENST00000458740.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225841.hg.1 |
NM_022460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200225841.hg.1 |
TC0200016640.hg.1 |
Clariom D TC |