Assay Details
| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003385.4 | NP_003376.2 | 2 - 3 | 540 | 95 | |
| GenBank mRNA | AB001104.1 | - | 2 - 3 | 295 | 95 | |
| AF039555.1 | - | 2 - 3 | 348 | 95 | ||
| BC022012.2 | - | 2 - 3 | 377 | 4823954 | 95 | |
| BI544338.1 | - | 2 - 3 | 541 | 95 | ||
| U14747.1 | - | 2 - 3 | 363 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000104.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02001217.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001217.hg.1 | TC02000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000483921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | ENST00000483921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001226.hg.1 | ENST00000295156.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001226.hg.1 | ENST00000404666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001226.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001226.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001226.hg.1 | TC02000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001221.hg.1 | ENST00000295156.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001221.hg.1 | ENST00000483921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001221.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001221.hg.1 | TC02000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001225.hg.1 | ENST00000295156.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001225.hg.1 | ENST00000404666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001225.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001225.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001225.hg.1 | TC02000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001218.hg.1 | ENST00000295156.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001218.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001218.hg.1 | TC02000104.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200146731.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146731.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146731.hg.1 | TC0200006831.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000483921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | ENST00000483921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006831.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146732.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146732.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146732.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146732.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146732.hg.1 | TC0200006831.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146735.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146735.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146735.hg.1 | ENST00000483921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146735.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146735.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146735.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146735.hg.1 | ENST00000483921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146735.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146735.hg.1 | TC0200006831.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146740.hg.1 | TC0200006831.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | ENST00000295156.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | ENST00000404666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | ENST00000406397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | ENST00000451533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | ENST00000457525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | NM_003385.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146741.hg.1 | TC0200006831.hg.1 | Clariom D TC |

VSNL1
Genomic Map