Assay Details
| Gene Symbol: AP4S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128126.2 | NP_001121598.1 | 3 - 4 | 611 | 109 | |
| NM_001254727.1 | NP_001241656.1 | 3 - 4 | 611 | 109 | ||
| NM_001254728.1 | NP_001241657.1 | 3 - 4 | 968 | 109 | ||
| NM_001254729.1 | NP_001241658.1 | 3 - 4 | 486 | 109 | ||
| NM_007077.4 | NP_009008.2 | 3 - 4 | 611 | 109 | ||
| XM_005267293.4 | XP_005267350.1 | 4 - 5 | 1054 | 109 | ||
| XM_011536371.2 | XP_011534673.1 | 4 - 5 | 624 | 109 | ||
| XM_011536372.2 | XP_011534674.1 | 3 - 4 | 1018 | 109 | ||
| GenBank mRNA | AB030654.1 | - | 3 - 4 | 431 | 109 | |
| AF155159.1 | - | 3 - 4 | 405 | 109 | ||
| AK304115.1 | - | 3 - 4 | 432 | 109 | ||
| AK316517.1 | - | 3 - 4 | 434 | 109 | ||
| BC001259.1 | - | 3 - 4 | 394 | 3454117 | 109 | |
| BT006701.1 | - | 2 - 3 | 222 | 109 | ||
| BX247969.1 | - | 3 - 4 | 413 | 109 | ||
| CR457100.1 | - | 2 - 3 | 222 | 109 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

AP4S1
Genomic Map