Assay Details
| Gene Symbol: LETMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243689.1 | NP_001230618.1 | 1 - 2 | 179 | 92 | |
| NM_001300765.1 | NP_001287694.1 | 1 - 2 | 179 | 92 | ||
| NM_015416.4 | NP_056231.3 | 1 - 2 | 179 | 92 | ||
| NR_045018.1 | 1 - 2 | 179 | 92 | |||
| NR_045019.1 | 1 - 2 | 179 | 92 | |||
| NR_045020.1 | 1 - 2 | 179 | 92 | |||
| XM_006719335.1 | XP_006719398.1 | 1 - 2 | 151 | 92 | ||
| XM_006719339.1 | XP_006719402.1 | 1 - 2 | 151 | 92 | ||
| XM_017019152.1 | XP_016874641.1 | 1 - 2 | 154 | 92 | ||
| XM_017019154.1 | XP_016874643.1 | 1 - 2 | 151 | 92 | ||
| XM_017019157.1 | XP_016874646.1 | 1 - 2 | 151 | 92 | ||
| XM_017019160.1 | XP_016874649.1 | 1 - 2 | 151 | 92 | ||
| XM_017019162.1 | XP_016874651.1 | 1 - 2 | 160 | 92 | ||
| XM_017019163.1 | XP_016874652.1 | 1 - 2 | 151 | 92 | ||
| XM_017019164.1 | XP_016874653.1 | 1 - 2 | 171 | 92 | ||
| XM_017019165.1 | XP_016874654.1 | 1 - 2 | 151 | 92 | ||
| XM_017019167.1 | XP_016874656.1 | 1 - 2 | 151 | 92 | ||
| XM_017019168.1 | XP_016874657.1 | 1 - 2 | 151 | 92 | ||
| GenBank mRNA | AF195651.2 | - | 1 - 2 | 151 | 92 | |
| AF329277.1 | - | 1 - 2 | 178 | 92 | ||
| AK000639.1 | - | 1 - 2 | 166 | 92 | ||
| AK057866.1 | - | 1 - 2 | 124 | 92 | ||
| AK127540.1 | - | 1 - 2 | 140 | 92 | ||
| AK225836.1 | - | 1 - 2 | 137 | 92 | ||
| AK301898.1 | - | 1 - 2 | 139 | 92 | ||
| AK304707.1 | - | 1 - 2 | 151 | 92 | ||
| AK315119.1 | - | 1 - 2 | 146 | 92 | ||
| AY259835.1 | - | 1 - 2 | 121 | 92 | ||
| AY259836.1 | - | 1 - 2 | 121 | 92 | ||
| BC064943.1 | - | 1 - 2 | 125 | 6009854 | 92 | |
| BM549678.1 | - | 151 | 92 | |||
| BX343550.2 | - | 1 - 2 | 120 | 92 | ||
| BX376670.2 | - | 1 - 2 | 120 | 92 | ||
| DC357852.1 | - | 1 - 2 | 179 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF329277.1, XM_006719339.1, XM_017019164.1, XM_017019165.1 transcripts.
Set Membership: |
|

LETMD1
Genomic Map