Assay Details
| Gene Symbol: KLHL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033328.1 | 1 - 2 | 410 | 85 | ||
| NR_033329.1 | 1 - 2 | 410 | 85 | |||
| XM_006715754.2 | XP_006715817.1 | 1 - 2 | 357 | 85 | ||
| XM_017012441.1 | XP_016867930.1 | 1 - 2 | 357 | 85 | ||
| GenBank mRNA | AK056390.1 | - | 1 - 2 | 361 | 85 | |
| BC009555.1 | - | 1 - 2 | 326 | 85 | ||
| EF560731.1 | - | 1 - 2 | 129 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000140.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK056390.1,BC009555.1,EF560731.1,XM_006715754.2,XM_017012441.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000414163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000414163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000414163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000459661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000459661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000459661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000459661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000459661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000477076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000477076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000477076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000477076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000477076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |

KLHL7
Genomic Map