Assay Details
| Gene Symbol: COX6B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057427.1 | - | 7 - 8 | 986 | 68 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000326529.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000586191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000587854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588572.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000588586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000589879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000590900.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | ENST00000593184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012051.hg.1 | NM_144613.4 | Refseq |

COX6B2
Genomic Map