Assay Details
| Gene Symbol: FAM84A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145175.2 | NP_660158.2 | 1 - 2 | 255 | 101 | |
| GenBank mRNA | AF370404.1 | - | 1 - 2 | 201 | 101 | |
| AJ417080.1 | - | 1 - 2 | 209 | 101 | ||
| AK092654.1 | - | 1 - 2 | 224 | 101 | ||
| AK314463.1 | - | 1 - 2 | 224 | 101 | ||
| BC029933.1 | - | 1 - 2 | 209 | 101 | ||
| BC052284.1 | - | 1 - 2 | 204 | 5454643 | 101 | |
| BI547868.1 | - | 1 - 2 | 260 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000087.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF370404.1,AJ417080.1,AK092654.1,AK314463.1,BC052284.1,NM_145175.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02001075.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001075.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001075.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001075.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001085.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001085.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001085.hg.1 | ENST00000359969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001085.hg.1 | ENST00000540701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001085.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001085.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001078.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001078.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001078.hg.1 | ENST00000540701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001078.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001078.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001083.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001083.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001083.hg.1 | ENST00000359969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001083.hg.1 | ENST00000540701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001083.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001083.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001089.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001089.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001089.hg.1 | ENST00000359969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001089.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001089.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001076.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001076.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001076.hg.1 | ENST00000359969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001076.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001076.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001068.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001068.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001068.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001068.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001069.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001069.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001069.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001069.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001077.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001077.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001077.hg.1 | ENST00000359969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001077.hg.1 | ENST00000540701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001077.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001077.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001081.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001081.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001081.hg.1 | ENST00000359969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001081.hg.1 | ENST00000540701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001081.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001081.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001080.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001080.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001080.hg.1 | ENST00000540701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001080.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001080.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001087.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001087.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001087.hg.1 | ENST00000359969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001087.hg.1 | ENST00000540701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001087.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001087.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001090.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001090.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001090.hg.1 | ENST00000359969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001090.hg.1 | ENST00000497769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001090.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001090.hg.1 | TC02000087.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200146441.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146441.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146441.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146441.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146441.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146441.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146441.hg.1 | TC0200006779.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146447.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146447.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146447.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146447.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146447.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146447.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146447.hg.1 | TC0200006779.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000464947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | ENST00000497769.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_001291410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006779.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146446.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146446.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146446.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146446.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146446.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146446.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146446.hg.1 | TC0200006779.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146440.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146440.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146440.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146440.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146440.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146440.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146440.hg.1 | TC0200006779.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146445.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146445.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146445.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146445.hg.1 | ENST00000295092.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146445.hg.1 | ENST00000331243.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146445.hg.1 | NM_145175.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146445.hg.1 | TC0200006779.hg.1 | Clariom D TC |

FAM84A
Genomic Map