| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10011249.hg.1 |
ENST00000368692.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10011249.hg.1 |
ENST00000368693.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10011249.hg.1 |
ENST00000368695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10011249.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10011249.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10011249.hg.1 |
TC10000917.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000417114.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000417114.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000417114.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000417114.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000417114.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000445510.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000445510.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000445510.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000445510.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000445510.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000494896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000494896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
ENST00000494896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368691.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000417114.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000417114.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000417114.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000417114.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000417114.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000445510.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000445510.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000445510.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000445510.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000445510.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000456942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000494896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000494896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
ENST00000494896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009248.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
ENST00000368693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
ENST00000368695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
ENST00000464130.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
ENST00000477963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
ENST00000492670.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
NM_145235.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000163452.hg.1 |
TC1000009248.hg.1 |
Clariom D TC |