Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006038.hg.1 |
NM_145274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006038.hg.1 |
TC17000500.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006039.hg.1 |
ENST00000301665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006039.hg.1 |
NM_145274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006039.hg.1 |
TC17000500.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006041.hg.1 |
ENST00000301665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006041.hg.1 |
ENST00000496847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006041.hg.1 |
NM_145274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006041.hg.1 |
TC17000500.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006040.hg.1 |
ENST00000301665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006040.hg.1 |
NM_145274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006040.hg.1 |
TC17000500.hg.1 |
HTA TC |