| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000808.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000808.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000808.hg.1 |
TC0X000060.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000243325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000243325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000243325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000243325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000243325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000243325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000243325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000243325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
ENST00000618931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_001195328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006656.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00145530.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00145530.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00145530.hg.1 |
ENST00000464506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00145530.hg.1 |
NM_004251.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00145530.hg.1 |
TC0X00006656.hg.1 |
Clariom D TC |