| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010299.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010299.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010299.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010299.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010299.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010299.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010299.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010299.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010299.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010299.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021472.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021472.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021472.hg.1 |
TC01001368.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021473.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021473.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021473.hg.1 |
TC01001368.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100010299.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010299.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010299.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010299.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010299.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010299.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010299.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010299.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010299.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010299.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175604.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175604.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175604.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175604.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175604.hg.1 |
TC0100010299.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100175605.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175605.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175605.hg.1 |
ENST00000302101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175605.hg.1 |
NM_005598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175605.hg.1 |
TC0100010299.hg.1 |
Clariom D TC |