Assay Details
| Gene Symbol: CBR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032783.4 | NP_116172.2 | 3 - 4 | 626 | 68 | |
| XM_005263315.2 | XP_005263372.1 | 3 - 4 | 593 | 68 | ||
| XM_005263316.3 | XP_005263373.1 | 3 - 4 | 593 | 68 | ||
| GenBank mRNA | AK027337.1 | - | 3 - 4 | 563 | 68 | |
| AK291756.1 | - | 3 - 4 | 563 | 68 | ||
| AL833393.1 | - | 3 - 4 | 566 | 68 | ||
| BC021973.2 | - | 3 - 4 | 545 | 3625459 | 68 | |
| BC033650.1 | - | 3 - 4 | 556 | 5574637 | 68 | |
| BQ230058.1 | - | 3 - 4 | 580 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001723.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000506808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000506808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000506808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000506808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000506808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000507752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000507752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000507752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000507752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000507752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023811.hg.1 | ENST00000306193.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023811.hg.1 | ENST00000504480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023811.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023811.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023811.hg.1 | TC04001723.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023813.hg.1 | ENST00000306193.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023813.hg.1 | ENST00000504480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023813.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023813.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023813.hg.1 | TC04001723.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400184810.hg.1 | TC0400012375.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400184814.hg.1 | TC0400012375.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000306193.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000504561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000506808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000506808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000506808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000506808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000506808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000507752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000507752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000507752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000507752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000507752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000509108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | ENST00000510042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | NM_032783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012375.hg.1 | NM_032783.4 | Refseq |

CBR4
Genomic Map