| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008853.hg.1 |
ENST00000353224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008853.hg.1 |
ENST00000378423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008853.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008853.hg.1 |
ENST00000439520.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008853.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008853.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008853.hg.1 |
TC20000615.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
TC2000008367.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158902.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000353224.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378423.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
ENST00000378429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_020341.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008367.hg.1 |
NM_177990.2 |
Refseq |