Assay Details
| Gene Symbol: TRMT10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134665.2 | NP_001128137.1 | 3 - 4 | 519 | 94 | |
| NM_001134666.2 | NP_001128138.1 | 3 - 4 | 508 | 94 | ||
| NM_152292.4 | NP_689505.1 | 3 - 4 | 664 | 94 | ||
| XM_005263352.3 | XP_005263409.1 | 3 - 4 | 498 | 94 | ||
| XM_006714417.2 | XP_006714480.1 | 3 - 4 | 540 | 94 | ||
| GenBank mRNA | AK000332.1 | - | 3 - 4 | 470 | 94 | |
| AK313548.1 | - | 3 - 4 | 494 | 94 | ||
| BC028373.2 | - | 3 - 4 | 664 | 4838346 | 94 | |
| EU668342.1 | - | 3 - 4 | 664 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001413.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000507394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000507394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000507394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000507394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000515831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000515831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | ENST00000515831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019746.hg.1 | ENST00000273962.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019746.hg.1 | ENST00000394876.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019746.hg.1 | ENST00000394877.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019746.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019746.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019746.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019746.hg.1 | TC04001413.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000507394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000507394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000507394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000507394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000515831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000515831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | ENST00000515831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011414.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000515831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000273962.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000394876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000394877.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000455368.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000514547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | ENST00000515831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | NM_001134665.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | NM_001134666.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | NM_152292.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178116.hg.1 | TC0400011414.hg.1 | Clariom D TC |

TRMT10A
Genomic Map