Assay Details
| Gene Symbol: GOT1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152413.2 | NP_689626.2 | 6 - 7 | 864 | 69 | |
| XM_005273399.3 | XP_005273456.1 | 6 - 7 | 1233 | 69 | ||
| XM_006716285.3 | XP_006716348.1 | 5 - 6 | 1144 | 69 | ||
| XM_011544391.2 | XP_011542693.1 | 3 - 4 | 320 | 69 | ||
| GenBank mRNA | BC029504.1 | - | 6 - 7 | 865 | 5273324 | 69 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001139.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26359096 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26359068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077152 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26359097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077145 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26359071 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077153 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077134 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26359090 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26359083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077143 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26359075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26359069 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077131 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26359073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077136 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077135 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077144 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06077142 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26359077 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 55448 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 300089 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 311240 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P9075 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3370449 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P9075 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P808720 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 4458 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 32169 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P109951 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P109951 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 53583 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42009 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P32247425 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P32247477 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P32247415 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P32247458 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P9075 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3370449 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P9075 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3370449 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P9075 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P808720 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P109951 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P109951 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P109951 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P9075 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P808720 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 76075 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 63982 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32247434 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18280755 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P848173 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P942758 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077149 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077159 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077155 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077153 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077136 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077125 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077131 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16087477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077127 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077148 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077134 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077140 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077143 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077145 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077129 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06077137 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18280755 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_152413_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8150246 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8150248 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8150247 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8150245 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8150244 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1553879_PM_a_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1553878_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11737542_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11737542_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014611.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014611.hg.1 | ENST00000518826.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014611.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014611.hg.1 | TC08001139.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000518826.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000518826.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000518826.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000518826.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000524298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000524298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000524298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000524298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | ENST00000524298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000518826.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000518826.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000518826.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000518826.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000524298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000524298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000524298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000524298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | ENST00000524298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010142.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168945.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168945.hg.1 | ENST00000518826.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168945.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168945.hg.1 | ENST00000307599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168945.hg.1 | ENST00000518826.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168945.hg.1 | NM_152413.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168945.hg.1 | TC0800010142.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1553879_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553879_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553878_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553878_PM_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_152413_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553878_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553879_a_at4 | - | - |
| Human Genome U95 Set | 76344_at | - | - |
| Human Genome U95 Set | 90303_at | - | - |
| Human Genome U95 Set | 90303_PM_at | - | - |
| Human Genome U95 Set | 76344_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035690 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035690 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 122107 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1778726 | BC029504.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778726 | BC029504.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1778726 | BC029504.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1778726 | BC029504.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1778726 | BC029504.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_22748874-S | BC029504.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_22748874-S | NM_152413.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_7880-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0005080022 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0005080022 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1778726 | BC029504.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1778726 | BC029504.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_7880 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_7880 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1778726 | BC029504.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778726 | BC029504.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778726 | BC029504.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1778726 | BC029504.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1778726 | NM_152413.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1778726 | BC029504.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1778726 | NM_152413.2 | human_transcriptome |

GOT1L1
Genomic Map