| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01011509.hg.1 |
ENST00000371076.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01011509.hg.1 |
ENST00000371077.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01011509.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01011509.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01011509.hg.1 |
TC01000724.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000608956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000608956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000608956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100161018.hg.1 |
TC0100008599.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371076.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000371077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000464349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000608956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
ENST00000611228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008599.hg.1 |
NM_152489.2 |
Refseq |