Assay Details
| Gene Symbol: LRRC47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020710.2 | NP_065761.1 | 4 - 5 | 1337 | 80 | |
| GenBank mRNA | AB033011.1 | - | 4 - 5 | 769 | 80 | |
| BC031301.1 | - | 4 - 5 | 1337 | 4330114 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002136.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032843.hg.1 | ENST00000378251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032843.hg.1 | ENST00000462356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032843.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032843.hg.1 | TC01002136.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032841.hg.1 | ENST00000378251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032841.hg.1 | ENST00000462356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032841.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032841.hg.1 | TC01002136.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100193268.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193268.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193268.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193268.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100193268.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193268.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193268.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193268.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100193268.hg.1 | TC0100012601.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012601.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100193270.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193270.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193270.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193270.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100193270.hg.1 | ENST00000378251.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193270.hg.1 | ENST00000462356.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193270.hg.1 | ENST00000479239.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100193270.hg.1 | NM_020710.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100193270.hg.1 | TC0100012601.hg.1 | Clariom D TC |

LRRC47
Genomic Map