Assay Details
| Gene Symbol: METTL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020961.3 | NP_066012.1 | 8 - 9 | 905 | 148 | |
| XM_005263166.4 | XP_005263223.1 | 8 - 9 | 900 | 148 | ||
| GenBank mRNA | AB046847.1 | - | 8 - 9 | 860 | 148 | |
| AK055555.1 | - | 8 - 9 | 883 | 148 | ||
| AK296056.1 | - | 6 - 7 | 525 | 148 | ||
| BC006565.2 | - | 8 - 9 | 856 | 3010654 | 148 | |
| BC007449.2 | - | 8 - 9 | 856 | 3010654 | 148 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000608.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008755.hg.1 | ENST00000388822.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008755.hg.1 | ENST00000506780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008755.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008755.hg.1 | TC04000608.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008757.hg.1 | ENST00000388822.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008757.hg.1 | ENST00000506780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008757.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008757.hg.1 | TC04000608.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158650.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158650.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158650.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158650.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158650.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158650.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158650.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158650.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158650.hg.1 | TC0400008547.hg.1 | Clariom D TC |

METTL14
Genomic Map