Assay Details
| Gene Symbol: PTRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC008849.1 | - | 1 - 2 | 85 | 4140111 | 149 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC008849.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185243.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185243.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185243.hg.1 | TC1700010723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700185243.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185243.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010723.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185250.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185250.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185250.hg.1 | TC1700010723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700185250.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185250.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185251.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185251.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185251.hg.1 | TC1700010723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700185251.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185251.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185242.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185242.hg.1 | NM_012232.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185242.hg.1 | TC1700010723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700185242.hg.1 | ENST00000357037.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185242.hg.1 | NM_012232.5 | Refseq |

PTRF
Genomic Map