Assay Details
| Gene Symbol: PHYHIPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143774.1 | NP_001137246.1 | 2 - 3 | 345 | 78 | |
| NM_032439.3 | NP_115815.2 | 2 - 3 | 565 | 78 | ||
| XM_011540275.2 | XP_011538577.1 | 3 - 4 | 627 | 78 | ||
| XM_011540276.2 | XP_011538578.1 | 2 - 3 | 1458 | 78 | ||
| XM_017016782.1 | XP_016872271.1 | 3 - 4 | 446 | 78 | ||
| XM_017016783.1 | XP_016872272.1 | 3 - 4 | 1549 | 78 | ||
| GenBank mRNA | AK054956.1 | - | 2 - 3 | 369 | 78 | |
| AL365474.1 | - | 2 - 3 | 345 | 78 | ||
| AL834339.1 | - | 2 - 3 | 566 | 78 | ||
| AY358162.1 | - | 1 - 2 | 195 | 78 | ||
| BC011268.2 | - | 2 - 3 | 404 | 4156912 | 78 | |
| CR749429.1 | - | 3 - 4 | 594 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000366.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10004217.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004217.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004217.hg.1 | TC10000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004221.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004221.hg.1 | ENST00000373880.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004221.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004221.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004221.hg.1 | TC10000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004222.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004222.hg.1 | ENST00000373880.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004222.hg.1 | ENST00000472199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004222.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004222.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004222.hg.1 | TC10000366.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000152430.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152430.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152430.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152430.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152430.hg.1 | TC1000007714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152433.hg.1 | TC1000007714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | ENST00000472199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | ENST00000373880.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | ENST00000472199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | ENST00000486074.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | NM_032439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152434.hg.1 | TC1000007714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000152429.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152429.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152429.hg.1 | ENST00000373878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152429.hg.1 | NM_001143774.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152429.hg.1 | TC1000007714.hg.1 | Clariom D TC |

PHYHIPL
Genomic Map