Assay Details
| Gene Symbol: SLC25A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000387.5 | NP_000378.1 | 8 - 9 | 1045 | 83 | |
| GenBank mRNA | AK301744.1 | - | 9 - 10 | 1006 | 83 | |
| AK312962.1 | - | 8 - 9 | 938 | 83 | ||
| BC001689.2 | - | 8 - 9 | 917 | 3050073 | 83 | |
| Y10319.1 | - | 8 - 9 | 908 | 83 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000319017.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000430379.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000440964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | ENST00000479050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011038.hg.1 | NM_000387.5 | Refseq |

SLC25A20
Genomic Map