Assay Details
| Gene Symbol: CD40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250.5 | NP_001241.1 | 6 - 7 | 656 | 64 | |
| NM_001322421.1 | NP_001309350.1 | 6 - 7 | 656 | 64 | ||
| NR_136327.1 | 5 - 6 | 562 | 64 | |||
| XM_017028137.1 | XP_016883626.1 | 6 - 7 | 665 | 64 | ||
| GenBank mRNA | AK222896.1 | - | 6 - 7 | 596 | 64 | |
| AY225405.1 | - | 6 - 7 | 566 | 64 | ||
| BC012419.1 | - | 6 - 7 | 623 | 3856903 | 64 | |
| BT019901.1 | - | 6 - 7 | 566 | 64 | ||
| KU177966.1 | - | 6 - 7 | 566 | 64 | ||
More Information
Set Membership: |
|

CD40
Genomic Map