Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303450.1 | NP_001290379.1 | 4 - 5 | 962 | 58 | |
| NM_021646.3 | NP_067678.1 | 4 - 5 | 962 | 58 | ||
| XM_005255243.3 | XP_005255300.1 | 3 - 4 | 928 | 58 | ||
| XM_011522453.2 | XP_011520755.1 | 4 - 5 | 963 | 58 | ||
| XM_011522454.2 | XP_011520756.1 | 3 - 4 | 717 | 58 | ||
| XM_017023121.1 | XP_016878610.1 | 3 - 4 | 455 | 58 | ||
| GenBank mRNA | AB011129.1 | - | 3 - 4 | 702 | 58 | |
| AK291792.1 | - | 4 - 5 | 906 | 58 | ||
| AK297993.1 | - | 4 - 5 | 961 | 58 | ||
| BC121013.1 | - | 3 - 4 | 688 | 40117042 | 58 | |
| BC121014.1 | - | 3 - 4 | 645 | 40117044 | 58 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000834.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16012121.hg.1 | ENST00000219478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16012121.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16012121.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16012121.hg.1 | TC16000834.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000585977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000585977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000585977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000585977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000588602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000588602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000588602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000588602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000588602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000588942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000588942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000592425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | ENST00000592425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000585977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000585977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000585977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000585977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000588602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000588602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000588602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000588602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000588602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000588942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000588942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000591026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000592425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | ENST00000592425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011476.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | NM_021646.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | TC1600011476.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | ENST00000219478.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | ENST00000545009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | ENST00000589422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | NM_001303450.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192569.hg.1 | NM_021646.3 | Refseq |

ZNF500
Genomic Map