Assay Details
| Gene Symbol: SNX13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015132.4 | NP_055947.1 | 1 - 2 | 227 | 79 | |
| XM_005249672.1 | XP_005249729.1 | 1 - 2 | 237 | 79 | ||
| XM_005249673.4 | XP_005249730.1 | 1 - 2 | 237 | 79 | ||
| XM_011515229.1 | XP_011513531.1 | 1 - 2 | 237 | 79 | ||
| XM_011515233.2 | XP_011513535.1 | 1 - 2 | 218 | 79 | ||
| XM_017011893.1 | XP_016867382.1 | 1 - 2 | 237 | 79 | ||
| XM_017011895.1 | XP_016867384.1 | 1 - 2 | 222 | 79 | ||
| XM_017011896.1 | XP_016867385.1 | 1 - 2 | 220 | 79 | ||
| GenBank mRNA | AB018256.1 | - | 1 - 2 | 187 | 79 | |
| AF420470.1 | - | 1 - 2 | 195 | 79 | ||
| AK315135.1 | - | 1 - 2 | 213 | 79 | ||
| AL353943.1 | - | 1 - 2 | 203 | 79 | ||
| BC045667.1 | - | 1 - 2 | 197 | 5298570 | 79 | |
| BG718588.1 | - | 1 - 2 | 221 | 79 | ||
| BX503974.1 | - | 1 - 2 | 228 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF420470.1, AK315135.1, AL353943.1, BC045667.1, NM_015132.4, XM_005249672.1, XM_005249673.4, XM_011515229.1, XM_011515233.2, XM_017011893.1, XM_017011895.1, XM_017011896.1 transcripts.
Set Membership: |
|

SNX13
Genomic Map