Assay Details
| Gene Symbol: ARFGEF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020340.4 | NP_065073.3 | 27 - 28 | 4642 | 90 | |
| GenBank mRNA | AB033070.2 | - | 18 - 19 | 3556 | 90 | |
| AB252196.1 | - | 27 - 28 | 4642 | 90 | ||
| AF413080.1 | - | 16 - 17 | 3250 | 90 | ||
| BC137174.1 | - | 25 - 26 | 4277 | 9021171 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001029.hg.1
Set Membership: |
|

ARFGEF3
Genomic Map