Assay Details
| Gene Symbol: DESI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015704.2 | NP_056519.1 | 5 - 6 | 632 | 112 | |
| GenBank mRNA | AF038183.1 | - | 5 - 6 | 590 | 112 | |
| BC093956.1 | - | 5 - 6 | 489 | 7939801 | 112 | |
| BC112179.1 | - | 5 - 6 | 489 | 8327647 | 112 | |
| BC171724.1 | - | 3 - 4 | 397 | 112 | ||
| CR456458.1 | - | 5 - 6 | 487 | 112 | ||
More Information
Set Membership: |
|

DESI1
Genomic Map