Assay Details
| Gene Symbol: DNAJC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015268.3 | NP_056083.3 | 52 - 53 | 6302 | 60 | |
| XM_005247245.4 | XP_005247302.1 | 53 - 54 | 6499 | 60 | ||
| XM_017006036.1 | XP_016861525.1 | 39 - 40 | 4759 | 60 | ||
| GenBank mRNA | AB014578.2 | - | 52 - 53 | 6351 | 60 | |
| AK023168.1 | - | 21 - 22 | 2730 | 60 | ||
| AK125330.1 | - | 29 - 30 | 3686 | 60 | ||
| AK127112.1 | - | 36 - 37 | 4390 | 60 | ||
| AY779857.1 | - | 52 - 53 | 6302 | 60 | ||
| BC000164.3 | - | 5 - 6 | 665 | 60 | ||
| BC009630.1 | - | 7 - 8 | 785 | 60 | ||
| BC040638.2 | - | 2 - 3 | 353 | 60 | ||
| BC065756.1 | - | 1 - 2 | 61 | 60 | ||
| BC151246.1 | - | 53 - 54 | 6372 | 9007477 | 60 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000715.hg.1
Set Membership: |
|

DNAJC13
Genomic Map