Assay Details
| Gene Symbol: PPP1R3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276318.1 | NP_001263247.1 | 3 - 4 | 1352 | 135 | |
| GenBank mRNA | AK024489.1 | - | 3 - 4 | 1352 | 135 | |
| AK091943.1 | - | 2 - 3 | 244 | 135 | ||
| BX161476.1 | - | 1 - 2 | 815 | 135 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BX161476.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 686979 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 519947 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 912347 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P428640 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P428640 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 14659 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 51530 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P57124595 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P428640 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P428640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624077 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167142 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16685470 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167151 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624102 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624106 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624100 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16685483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624096 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624087 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624066 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16685470 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167142 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624077 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624096 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624087 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624100 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624066 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624106 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167151 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624102 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29624085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16685483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09167126 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P428640 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P19978874 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P428640 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19978874 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02875007 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167176 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167105 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167185 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167151 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167160 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167132 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167157 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167101 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167091 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167163 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685458 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685490 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167125 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167224 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167166 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167202 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685483 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685481 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167096 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167187 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167120 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167206 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685486 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167208 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167169 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167114 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167184 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167226 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167130 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685478 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167085 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167148 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167199 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167136 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167177 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167103 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167088 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167190 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167193 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167174 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167172 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167094 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167118 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167140 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167126 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685449 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167195 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167164 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167210 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167134 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167084 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167099 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167197 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167117 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167180 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685452 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167155 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167213 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167204 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685474 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685467 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16685457 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167218 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167145 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167123 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09167141 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19978874 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02874996 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7977961 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7977963 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7977960 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7977964 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7977962 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7977955 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 229001_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 227412_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 227409_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11751738_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11751738_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 229001_at | - | - |
| Human Genome U133 Plus 2.0 Array | 227412_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 227412_at | - | - |
| Human Genome U133 Plus 2.0 Array | 227409_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 229001_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 227409_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229001_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227412_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227409_at4 | - | - |
| Human Genome U95 Set | 45163_PM_at | - | - |
| Human Genome U95 Set | 45163_at | - | - |
| Human Genome U95 Set | 64205_PM_at | - | - |
| Human Genome U95 Set | 61974_PM_at | - | - |
| Human Genome U95 Set | 64205_at | - | - |
| Human Genome U95 Set | 61974_at | - | - |
| Human Genome U95 Set | 73322_at | - | - |
| Human Genome U95 Set | 73322_PM_at | - | - |
| Human Genome U95 Set | 77259_at | - | - |
| Human Genome U95 Set | 77259_PM_at | - | - |
| Human Genome U133 Set | 229001_at | - | - |
| Human Genome U133 Set | 227412_PM_at | - | - |
| Human Genome U133 Set | 227412_at | - | - |
| Human Genome U133 Set | 227409_PM_at | - | - |
| Human Genome U133 Set | 227409_at | - | - |
| Human Genome U133 Set | 229001_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050618 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050618 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 153038 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 206803 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1735064 | CR619320.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1735064 | NR_026862.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1735064 | AK024489.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1735064 | AW663470.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1735064 | CR619320.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1735064 | NR_026862.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1735064 | AK024489.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1735064 | AW663470.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1735064 | CR619320.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1735064 | NR_026862.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1735064 | AK024489.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1735064 | AW663470.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1735064 | CR619320.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1735064 | NR_026862.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1735064 | AK024489.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1735064 | AW663470.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1735064 | CR619320.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1735064 | NR_026862.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1735064 | AK024489.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1735064 | AW663470.1 | human_transcriptome |

PPP1R3E
Genomic Map