Assay Details
| Gene Symbol: SNAPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006049.2 | NP_006040.1 | 1 - 2 | 168 | 101 | |
| GenBank mRNA | AF093593.1 | - | 1 - 2 | 91 | 101 | |
| AK024763.1 | - | 1 - 2 | 140 | 101 | ||
| AK312018.1 | - | 1 - 2 | 140 | 101 | ||
| CD685245.1 | - | 1 - 2 | 205 | 101 | ||
| CN347010.1 | - | 1 - 2 | 90 | 101 | ||
| DB037177.1 | - | 1 - 2 | 100 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001578.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015186.hg.1 | ENST00000316634.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015186.hg.1 | ENST00000395589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015186.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015186.hg.1 | TC15001578.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015189.hg.1 | ENST00000316634.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015189.hg.1 | ENST00000395589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015189.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015189.hg.1 | TC15001578.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015187.hg.1 | ENST00000307979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015187.hg.1 | ENST00000316634.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015187.hg.1 | ENST00000395589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015187.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015187.hg.1 | TC15001578.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | TC1500010883.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184997.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500185005.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500185005.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500185005.hg.1 | TC1500010883.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500185005.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500185005.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |

SNAPC5
Genomic Map