| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038264.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038264.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038264.hg.1 |
TC03002161.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202447.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202447.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202447.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202447.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202447.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202447.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202447.hg.1 |
TC0300013703.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000318037.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
ENST00000437070.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013703.hg.1 |
NM_152617.3 |
Refseq |