Assay Details
| Gene Symbol: GLT8D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316967.1 | NP_001303896.1 | 8 - 9 | 1252 | 69 | |
| NM_031302.4 | NP_112592.1 | 9 - 10 | 1387 | 69 | ||
| XM_011538793.2 | XP_011537095.1 | 9 - 10 | 1622 | 69 | ||
| XM_017019999.1 | XP_016875488.1 | 10 - 11 | 1703 | 69 | ||
| GenBank mRNA | AJ271711.1 | - | 6 - 7 | 601 | 69 | |
| AK027310.1 | - | 7 - 8 | 701 | 69 | ||
| AK056056.1 | - | 8 - 9 | 1258 | 69 | ||
| AY358763.1 | - | 8 - 9 | 791 | 69 | ||
| BC022343.1 | - | 8 - 9 | 1008 | 4248949 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000546851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000552572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000552572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | ENST00000552572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000546851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000547583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000550816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000552572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000552572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | ENST00000552572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011752.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191131.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191131.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191131.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191131.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191131.hg.1 | ENST00000360814.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191131.hg.1 | ENST00000546436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191131.hg.1 | ENST00000548660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191131.hg.1 | NM_031302.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191131.hg.1 | TC1200011752.hg.1 | Clariom D TC |

GLT8D2
Genomic Map