Assay Details
| Gene Symbol: CHPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284295.1 | NP_001271224.1 | 1 - 2 | 325 | 116 | |
| GenBank mRNA | AK056331.1 | - | 1 - 2 | 325 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK056331.1, NM_001284295.1 transcripts.
Set Membership: |
|

CHPF2
Genomic Map