Assay Details
| Gene Symbol: ANKRD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144994.7 | NP_659431.5 | 5 - 6 | 556 | 89 | |
| GenBank mRNA | AF492401.1 | - | 5 - 6 | 532 | 89 | |
| AJ315766.1 | - | 4 - 5 | 500 | 89 | ||
| AK057011.1 | - | 5 - 6 | 556 | 89 | ||
| AY196212.1 | - | 5 - 6 | 540 | 89 | ||
| BC107056.2 | - | 5 - 6 | 551 | 40005436 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02005037.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034246.hg.1 | ENST00000318357.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034246.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034246.hg.1 | ENST00000418232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034246.hg.1 | ENST00000476975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034246.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02034246.hg.1 | TC02005037.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200227484.hg.1 | TC0200016712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000318357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000331001.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000418232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | ENST00000482175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016712.hg.1 | NM_144994.7 | Refseq |

ANKRD23
Genomic Map