Assay Details
| Gene Symbol: ENPP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153343.3 | NP_699174.1 | 4 - 5 | 815 | 89 | |
| GenBank mRNA | AK057370.1 | - | 4 - 5 | 760 | 89 | |
| AY358676.1 | - | 4 - 5 | 756 | 89 | ||
| BC035035.2 | - | 4 - 5 | 720 | 5259487 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001800.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04024539.hg.1 | ENST00000296741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024539.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024539.hg.1 | TC04001800.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000505644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000505644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024540.hg.1 | ENST00000296741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024540.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024540.hg.1 | TC04001800.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400186165.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186165.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186165.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186165.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186165.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186165.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186165.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186165.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186165.hg.1 | TC0400012595.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400186166.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186166.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186166.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186166.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186166.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186166.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186166.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186166.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186166.hg.1 | TC0400012595.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000505644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000505644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |

ENPP6
Genomic Map