Assay Details
| Gene Symbol: GRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002086.4 | NP_002077.1 | 4 - 5 | 653 | 83 | |
| GenBank mRNA | AF498925.1 | - | 3 - 4 | 296 | 83 | |
| AK091010.1 | - | 4 - 5 | 1081 | 83 | ||
| AK315565.1 | - | 4 - 5 | 649 | 83 | ||
| BC000631.2 | - | 4 - 5 | 598 | 3345524 | 83 | |
| BI459414.1 | - | 4 - 5 | 660 | 83 | ||
| CR450363.1 | - | 3 - 4 | 296 | 83 | ||
| CR541942.1 | - | 3 - 4 | 296 | 83 | ||
| M96995.1 | - | 3 - 4 | 374 | 83 | ||
| X62852.1 | - | 1 - 2 | 125 | 83 | ||
More Information
Set Membership: |
|

GRB2
Genomic Map