Assay Details
| Gene Symbol: CCDC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286243.1 | NP_001273172.1 | 9 - 10 | 1483 | 68 | |
| NM_001286244.1 | NP_001273173.1 | 8 - 9 | 1313 | 68 | ||
| NM_152591.2 | NP_689804.1 | 10 - 11 | 1588 | 68 | ||
| XM_006719262.1 | XP_006719325.1 | 11 - 12 | 1780 | 68 | ||
| XM_006719263.2 | XP_006719326.1 | 10 - 11 | 1541 | 68 | ||
| XM_011537999.1 | XP_011536301.1 | 11 - 12 | 1831 | 68 | ||
| XM_011538001.2 | XP_011536303.1 | 11 - 12 | 1823 | 68 | ||
| GenBank mRNA | AK093162.1 | - | 10 - 11 | 1541 | 68 | |
| AK302207.1 | - | 9 - 10 | 1483 | 68 | ||
| BC044815.1 | - | 10 - 11 | 1506 | 68 | ||
| BC064580.1 | - | 8 - 9 | 1246 | 68 | ||
| BC073147.1 | - | 9 - 10 | 1397 | 5742508 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000861.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011321.hg.1 | ENST00000308208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011321.hg.1 | ENST00000545036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011321.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011321.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011321.hg.1 | TC12000861.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200165715.hg.1 | TC1200008855.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000308208.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000545036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000550317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | ENST00000552694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286243.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_001286244.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008855.hg.1 | NM_152591.2 | Refseq |

CCDC63
Genomic Map