Assay Details
| Gene Symbol: SLC41A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173854.5 | NP_776253.3 | 2 - 3 | 1404 | 60 | |
| XM_005245069.2 | XP_005245126.1 | 2 - 3 | 1080 | 60 | ||
| GenBank mRNA | AJ514402.1 | - | 1 - 2 | 491 | 60 | |
| AK128069.1 | - | 1 - 2 | 657 | 60 | ||
| AK314732.1 | - | 1 - 2 | 562 | 60 | ||
| AL831974.1 | - | 2 - 3 | 1240 | 60 | ||
| AL832001.1 | - | 2 - 3 | 1240 | 60 | ||
| AL832362.1 | - | 2 - 3 | 1240 | 60 | ||
| BC136303.1 | - | 1 - 2 | 885 | 9020290 | 60 | |
| BX641054.1 | - | 2 - 3 | 1242 | 60 | ||
| BX648979.1 | - | 2 - 3 | 1278 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003754.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular magnesium ion homeostasis
magnesium ion transport
sodium ion transmembrane transport
transmembrane transport
divalent metal ion transport
cellular response to magnesium ion
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity
substrate-specific transporter activity
magnesium:sodium antiporter activity
divalent inorganic cation transmembrane transporter activity
cellular magnesium ion homeostasis
magnesium ion transport
sodium ion transmembrane transport
transmembrane transport
divalent metal ion transport
cellular response to magnesium ion
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity
substrate-specific transporter activity
magnesium:sodium antiporter activity
divalent inorganic cation transmembrane transporter activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01057583.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057583.hg.1 | ENST00000468057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057583.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057583.hg.1 | TC01003754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057589.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057589.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057589.hg.1 | TC01003754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057587.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057587.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057587.hg.1 | TC01003754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057586.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057586.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057586.hg.1 | TC01003754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057582.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057582.hg.1 | ENST00000468057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057582.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057582.hg.1 | TC01003754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057588.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057588.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057588.hg.1 | TC01003754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057584.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057584.hg.1 | ENST00000468057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057584.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057584.hg.1 | TC01003754.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100229441.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229441.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229441.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229441.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229441.hg.1 | TC0100017079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100229435.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229435.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229435.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229435.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229435.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229435.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229435.hg.1 | TC0100017079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100229438.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229438.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229438.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229438.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229438.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229438.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229438.hg.1 | TC0100017079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100229440.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229440.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229440.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229440.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229440.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229440.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229440.hg.1 | TC0100017079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000468057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | ENST00000484228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017079.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229437.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229437.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229437.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229437.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229437.hg.1 | TC0100017079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100229439.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229439.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229439.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229439.hg.1 | ENST00000367137.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229439.hg.1 | ENST00000484000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229439.hg.1 | NM_173854.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229439.hg.1 | TC0100017079.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 220276 | - | - |

SLC41A1
Genomic Map